3-Cyanoumbelliferone ≥95%

Supplier: AAT Bioquest
Warning

40
76480-480EA 96.26 USD
76480-480
3-Cyanoumbelliferone ≥95%
3-Cyanoumbelliferone

3-Cyano-7-hydroxycoumarin is used as a calibration standard for 3-cyano-7-hydroxycoumarin-based enzyme substrates.


  • pH-dependent fluorescence

Formula: C₁₀H₅NO₃
MDL Number: MFCD00037480
CAS Number: 19088-73-4
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR