Linearity FD General Chemistry Panel 1 for Abbott Systems, AUDIT Microcontrols
Supplier: AUDIT MICROCONTROLS
K940M-5
76480-188EA 345.25 USD
76480-188
Linearity FD General Chemistry Panel 1 for Abbott Systems, AUDIT Microcontrols
Controls and Calibration Verification Materials
It is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for BUN, calcium, chloride, CO₂, creatinine, glucose, iron, lactate, magnesium, phosphate, potassium, sodium, and uric acid.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.