Linearity FD iPTH for Ortho Vitros, AUDIT MircoControls
Supplier: AUDIT MICROCONTROLS
K904M-5
76479-170EA 470.43 USD
76479-170
Linearity FD iPTH for Ortho Vitros, AUDIT MircoControls
Controls and Calibration Verification Materials
Intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for iPTH.
The Linearity FD iPTH for Ortho Vitros is a five level linearity set. Each level contains the following analytes: iPTH. These five levels demonstrate a linear relationship to each other for their respective analytes.