Hypersil™ Chiral-IXT HPLC Column
Supplier: Thermo Scientific
42305-054630
76477-182EA 1876.6 USD
76477-182
Hypersil™ Chiral-IXT HPLC Column
Chromatography Columns
Separate a wide range of enantiomers with Thermo Scientific™ Hypersil™ Chiral HPLC columns, designed for normal-phase separations of neutral, acidic, and basic racemates.
These chiral columns help meet your purification needs by resolving racemic mixtures and potential contaminants so you can be confident in the purity of your target molecule.
To maximize screening efficiency, these columns can be ordered in kits containing one column in each of our chiral phases. All columns are available in analytical and preparative scaled formats.