Standard Sample Pan Crimper Press, PerkinElmer
Supplier: PERKINELMER U.S. LLC
02190048
101677-284EA 2997.33 USD
101677-284 76474-986
Standard Sample Pan Crimper Press, PerkinElmer
Crimpers, Cappers and Decappers
Used to crimp covers on standard DSC pans of aluminum, gold and copper.
  • Design incorporates a replaceable crimper head
  • Compatible with DSC