Thermomechanical Analyzer, TMA 4000, PerkinElmer
Supplier: PerkinElmer
L8040000
76474-916EA 45906.72 USD
76474-916
Thermomechanical Analyzer, TMA 4000, PerkinElmer
Analyzers Texture Analysers
The TMA 4000 is a uniquely designed thermomechanical analyzer (TMA) designed for optimal performance in measuring the coefficient of thermal expansion (CTE) in materials.
  • Temperature range –80 to 800 °C
  • Thermal analysis
  • Not portable
This rugged and easy-to-use design uses both electronics and a unique Archimedean float system to control the probe, allowing accurate determination of CTE and of transitions. In these times of budgetary constraints and changes in material requirements driven by regulations like RoHS, understanding how a material changes with temperature is more important than ever. TMA 4000 provides a tool designed to collect this data. Data can be collected from samples in many forms with the TMA 4000's range of probes including expansion, compression, flexure, extension, and volumetric (dilatometry).
Ordering information: One year warranty.