Intracoolers, PerkinElmer
Supplier: PerkinElmer
N5340040
101688-076EA 7628.5 USD
101688-076 76474-858
Intracoolers, PerkinElmer
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
The Intracooler Series cool the sample holder enclosure block of the DSC instruments.
  • It is designed for use with Differential scanning calorimeters (DSC)
They are closed-loop circulating heat exchangers with a compact mechanical refrigeration unit. They are an excellent economical option for laboratories requiring low temperatures or fast cooling rates, but with limited access to liquid N2. The Intracooler II series has a dual stage heat exchanger with a temperature range that encompasses most DSC 4000/6000 applications work.