PolyScience Chiller, PerkinElmer
Supplier: PerkinElmer
N5370220
76474-846EA 6229.89 USD
76474-846
PolyScience Chiller, PerkinElmer
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The chiller provides dependable temperature control for the TGA 4000/8000 instruments.
  • Depending on the coolant used, the fluid bath can reach a maximum low temperature of:
  • Distilled H₂O: 10 °C
  • Ethylene glycol (30%/H₂O 70%): 0 °C
  • Ethylene glycol (50%/H₂O 50%): −20 °C
When combined with the sub-ambient cooling kit (10138-634), the Chiller enables the TGA unit to operate under sub-ambient to elevated temperatures (−5 to +200 °C). It also supports faster furnace cool down at the end of experiments using raised temperatures. Reservoir capacity of 7 L.