PYRIS 6 DSC Intracoolers (115 V, 50/60 Hz), PerkinElmer
Supplier: PerkinElmer
N5374098
101688-074EA 7315 USD
101688-074 76474-844
PYRIS 6 DSC Intracoolers (115 V, 50/60 Hz), PerkinElmer
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
Intracoolers from PerkinElmer are subambient devices that automatically control the flow of coolant through a circulating chamber attached to the PYRIS 1 DSC, PYRIS 6 DSC, DSC 7, TMA 7, or DMA 7 analyzers.
They are either single or dual-stage mechanical compression refrigeration units with cylindrical expansion chamber and special bolts and insulation for mounting directly on the sample holder block of the PYRIS 1 DSC, PYRIS 6 DSC, and DSC 7, or the furnace assembly of the DMA 7 or TMA 7. Intracooler 1 permits operation of all analyzers from −30 °C; the Intracooler 2 from −65 °C.