Warmbier Metriso® B530 ESD Audit Kit, Transforming Technologies
Supplier: Transforming Technologies
7100.B530.MK
76474-614EA 4910.16 USD
76474-614
Warmbier Metriso® B530 ESD Audit Kit, Transforming Technologies
Resistance Testers
Kit includes the relevant test instruments and equipment needed to perform ESD audits.
  • Measure electrostatic fields
  • Measure surface resistance
  • Analyze ionizers performance
  • Evaluate packaging materials
  • Evaluate garments
  • Measure flooring, work surfaces and furniture resistance
  • Measure temperature and relative humidity
The Metriso® B530 ESD Audit Kit includes the relevant test instruments and equipment needed to perform ESD audits according to ANSI ESD.2020 and IEC 61340-5-1 and for qualification/verification of ESD control elements. The kit is built around the Metriso® B530 digital high resistance meter. It can take resistance measurements of 1 to 199 kΩ.