Stuart® Hotplates, Undergrad Series, Cole Parmer
Supplier: Antyila Scientific
04807-51 04807-53
76474-318EA 452.4 USD
76474-318 76474-320
Stuart® Hotplates, Undergrad Series, Cole Parmer
Heaters Hot Plates
Get accurate surface temperature in real time.
  • BioCote® glaze for antimicrobial protection
  • Compact, stackable design takes up less benchtop and storage space
  • Hot warning light remains on even when unplugged
  • Tough aluminum casing for longevity
  • Designed to prevent accidental spillage on control panel
  • IP 32 rated for water and dust resistance
Designed with microprocessor-controlled dual thermocouples, these hotplates offer accurate temperature control and protection from overheating. A colored LED indicator surrounds the heat control knob illuminating the actual surface temperature in real time. The square plate design allows flexibility of vessel sizes and combinations. Up to four 250 ml beakers can easily fit on the surface at the same time.
To keep the user safe, a 'hot' warning light flashes whenever the plate surface temperature is above 50 °C and continues to flash until the temperature drops below 50 °C, even when unplugged. The units also feature BioCote® antimicrobial protection which supports a safer and more hygienic laboratory environment, while reducing risk of cross-contamination in the laboratory.
All models include an integral fitting for a retort rod and allow the retort base to slide underneath. For convenience and power saving, the units are built with a standard mains switch.
Available with either chemically resistant glass ceramic top plate that stays cool around the edges to help prevent burns or coated aluminum top plate for rapid heating and better temperature uniformity. Compatible with the Stuart® SCT2 temperature controller.
Dimensions (L×W×H): 248×172×122 mm (9.76×6.77×4.8")
Certifications: IP protection class according to DIN EN 60529: IP 32.