Quidel QuickVue RSV Test, NDC
Supplier: Healthcare Products
QUI 20193
76474-228KT 380.22 USD
76474-228
Quidel QuickVue RSV Test, NDC
Clinical Diagnostic Supplies Clinical Reagents and Kits
The QuickVue RSV Test is a dipstick immunoassay that detects respiratory syncytial virus (RSV) antigen (viral fusion protein) directly from nasopharyngeal swab, nasopharyngeal aspirate, or nasal/nasopharyngeal wash specimens for symptomatic pediatric patients (18 years of age and younger).
  • Processing time of 15 minutes or less
  • CLIA complexity is waved