FLM300 Fluorophos ALP Test System, Advanced Instruments
Supplier: ADVANCED INSTRUMENTS INC.
FLM300
76474-156EA 14148.72 USD
76474-156
FLM300 Fluorophos ALP Test System, Advanced Instruments
Assays Enzyme Assays
The Fluorophos ALP assay is a rapid test for verifying proper pasteurization of dairy products.
  • Fast and accurate results
  • Globally proven
  • Reduced production liability
Alkaline Phosphatase (ALP), found naturally in raw milk, is destroyed at temperatures slightly above the temperature necessary to destroy microorganisms pathogenic to humans. Measurement of residual phosphatase after pasteurization offers a means of verifying the products are heated to the proper temperatures and are not contaminated with raw milk. The Fluorophos ALP assay measures the ALP activity in dairy products, allowing the user to monitor the completeness of pasteurization and detect raw milk contamination.