VWR® Drying Rack Set, Wall/Bench Mount
Supplier: VWR International
76474-088
76474-088EA 251.33 USD
76474-088
VWR® Drying Rack Set, Wall/Bench Mount
Racks Draining Racks
Versatile rack made of durable, easy to clean PS.
  • Features channels designed for more configuration and to protect against breakage
  • One of a kind drying baskets to collect small items
  • Unique peg angles allow for better hold - especially large, heavy pieces
  • SureGrip hook securely grips glassware into place
The VWR® drying rack can be used on counters with an integrated hinged stand, or walls with included guide and hardware. The rack can hold up to 77 pounds.
Delivery information: Includes 72 pegs (36 small, and 36 large), 2 drying baskets, and 1 drainage tube.