Rapid Response™ hCG Pregnancy Test Cassette (Urine/Serum), BTNX
Supplier: BTNX INC.
HCG-3C25
76473-492KT
33.99
USD
76473-492
Rapid Response™ hCG Pregnancy Test Cassette (Urine/Serum), BTNX
Clinical Diagnostic Supplies
Clinical Reagents and Kits
The Rapid Response® hCG Pregnancy combo test cassette is a rapid, qualitative test for the determination of human chorionic gonadotropin in human urine and serum specimens, to aid in the early detection of pregnancy.
- Format: Cassette
- Specificity: Human Chorionic Gonadotropin
- Accuracy: >99.9%
- Time to result: 5 minutes
- Storage condition: 2 -to 30 °C (36 to 86 °F)
- Test principle: Lateral flow immunoassay
Delivery information: Includes test cassettes, disposable pipettes and product insert.
Caution: This test is intended for in vitro diagnostic use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...