Rapid Response™ hCG Pregnancy Midstream Test Cassette, BTNX
Supplier: BTNX INC.
HCG-1M1
76473-486KT
3.4
USD
76473-486
Rapid Response™ hCG Pregnancy Midstream Test Cassette, BTNX
Clinical Diagnostic Supplies
Clinical Reagents and Kits
The Rapid Response® hCG Pregnancy midstream test is a rapid, self-performing, qualitative test for the determination of human chorionic gonadotropin in human urine specimens, to aid in the early detection of pregnancy.
- Sample: Urine
- Format: Midstream
- Specificity: Human Chorionic Gonadotropin
- Time to result: 3 minutes
- Storage condition: 36 to 86 °F (2 to 30 °C)
- Test principle: Lateral flow immunoassay
Delivery information: Includes midstream test, product insert.
Caution: This test is intended for in vitro diagnostic use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...