Rapid Response™ ETG Test Panel (Urine), BTNX
Supplier: BTNX INC.
ETG-1P9-25
76473-470KT 90.64 USD
76473-470
Rapid Response™ ETG Test Panel (Urine), BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response® Test panel that detects in urine ethyl glucuronide (ETG) at cut off of 500 ng/ml.
  • Parameter: ETG
  • Calibrator: Ethyl Glucuronide
  • Cut-off (ng/ml): 500
Delivery information: Includes test panels and product insert.