Rapid Response™ Liquid Diptube Urine Controls, BTNX
Supplier: BTNX INC.
AUCT-1V1
76473-394EA 101.96 USD
76473-394
Rapid Response™ Liquid Diptube Urine Controls, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response™ Liquid Diptube Urine Controls are available in two levels and are intended for use in validating the visual and analyzer reading of urinalysis, to ensure the consistent performance of Rapid Response® Urinalysis Reagent Strips and Rapid Response® U120 Urine Analyzers.
  • Available in two levels
They are intended for in vitro diagnostic use only. For visual reading, test results are interpreted by direct comparison of the color blocks printed on the canister label. For Analyzer reading, refer to the Instruction Manual for details.
Delivery information: Contents include 12 ml Level 1 control tube, 12 ml Level 2 control tube, and product insert.
Caution: This test is intended for in vitro diagnostic use only.