Rapid Response® Plastic Collection Cup, BTNX
Supplier: BTNX INC.
02SPC-14
76473-384BG 33.61 USD
76473-384
Rapid Response® Plastic Collection Cup, BTNX
Sample Containers General Purpose Containers
The Rapid Response® Plastic collection cup is a non sterile cup made with polypropylene allowing for clarity and durability and is ideal for sample collection.
  • 90 ml volume
  • 45 mm diameter
  • Screw-on cap
The cup includes graduations on the cup exterior for easy reading and made with polypropylene material for clarity and durability when reviewing samples. The compact size is ideal for sample testing with strips, cassettes, and 1 - 12 panel drug test.
Delivery information: Includes collection cups.