USP and EP Vis Standards Set for Spectrophotometers, Thermo Scientific
Supplier: THERMO ELECTRON NORTH AMERICA LLC
222-327300
76472-754EA
3650
USD
76472-754
USP and EP Vis Standards Set for Spectrophotometers, Thermo Scientific
Standards
Chromatography Standards
Reference Materials to quality performance verification (PV) tests in UV-Vis region. A set of filter standards for pharmacopeia compliance support.
- Wavelength accuracy
- Photometric accuracy
Wavelength Accuracy: Didymium glass filter
Photometric Accuracy: 4 neutral density filters calibrated for absorbance at 440, 465, 546.1, 590 and 635 nm. Absorbance values approximately 0.5, 1.0, 1.5 and 2.0.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)