USP and EP UV Standards Set for Spectrophotometers, Thermo Scientific
Supplier: THERMO ELECTRON NORTH AMERICA LLC
Reference Materials to quality performance verification (PV) tests in UV region. A set of solutions for pharmacopeia compliance support.
- Wavelength accuracy
- Absorbance accuracy
- Stray light
- Resolution
Wavelength Accuracy: Calibrated holmium oxide solution for wavelength accuracy from 241 nm to 641 nm
Photometric Accuracy: Calibrated 60 mg/L, 80 mg/L and 140 mg/L potassium dichromate with blank: UV absorbance accuracy to 2A
Stray Light: Certified potassium chloride, sodium iodide, and sodium nitrate for stray light measurements
Resolution: Certified toluene in hexane solution with hexane blank for resolution (spectral bandwidth) testing
Calibrated for use with instruments with SBW of < 3 nm.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...