Melting Point Apparatus, METTLER TOLEDO

Supplier: Mettler Toledo
30578102 30578100 30578101
76471-014EA 19030 USD
76471-014 76472-610 76472-612
Melting Point Apparatus, METTLER TOLEDO
Melting Point Apparatus
Melting point determination has never been so easy - with simple operating routines and an intuitive color touch screen, measurements can be performed quickly.

  • Everything needed for the simple, automatic determination of melting point
  • One Click™ melting point determination
  • Widescreen display with color touch screen
  • Color video recording
  • Intensity curves and results can be viewed directly on the color touch screen
  • Capability to export data as PDF

This melting point systems offer simultaneous determination of samples, statistical analysis, and printable results. The outstanding temperature-accuracy specifications and the compliance with current standards such as Ph. Eur., USP and others ensure accurate results for melting point and melting range determination up to 300 °C.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR