COWLES Impeller Guard, Safety Mechanism for Hi-Shear Impellers, MorehouseCowles
Supplier: MorehouseCowles
488972
76472-542EA 158.12 USD
76472-542
COWLES Impeller Guard, Safety Mechanism for Hi-Shear Impellers, MorehouseCowles
Homogenizers Dispersing Tools
The COWLES impeller guard from MorehouseCowles, puts a focus on safety. The guard covers the COWLES hi-shear impeller allowing safe and easy handling of the blade. It makes in-tank maintenance and cleaning much easier by eliminating the risk of cuts and prevents injury from accidental bumping of the exposed impeller. Changing impeller blades is now possible without handling sharp edges.
  • Light weight
  • Adaptable easy-to-use design
  • Modular design is convenient for any impeller size
  • Resistant to many common chemicals for easy cleaning
  • Lock-out, tag-out adapter is also available
The COWLES Impeller Guard includes 10 links that covers a 6" diameter impeller.
Accessories information: Additional individual links can be purchased separately to fit most diameter sizes. See the data sheet for details on the number of links required for standard diameters. Replacement latching mechanisms and locks for lock-out tag-out procedures (pad lock not included) can also be purchased separately.