Corning® X-MINI® Pressor, Corning
Supplier: Corning
6915
76470-988CS 3156.56 USD
76470-988
Corning® X-MINI® Pressor, Corning
Bioreactors
The Corning X-MINI Pressor is designed to be used exclusively with the Corning X-MINI Selection Kits.
The Corning X-MINI Pressor allows for the degassing of microbubble-targeted cells. It accommodates a capped 10 ml syringe.
The pressure generated using the X-MINI Pressor is sufficient to isolate up to 1×10⁷ cells, and one syringe can process up to 3 ml of cell suspension at a time.