VWR® Alliance Protector Sleeves, Made in USA
Supplier: VWR International
76470-788 76470-702
76470-788CS 714.58 USD
76470-788 76470-702
VWR® Alliance Protector Sleeves, Made in USA
Disposable Garments
VWR® Made in USA Sleeves are an ideal option for single-use needs. Made with the specially formulated Alliance breathable microporous fabric.
  • Made with Alliance breathable microporous fabric
  • Available with serged or bound seams
  • Produced with latex-free elastic
  • Made in USA
A key part of the VWR® Made in USA PPE line, these cleanroom sleeves are 20" in length to provide a comfortable fit while meeting the requirements of many controlled environments.
The Alliance fabric is designed to lower particulate levels by providing extremely smooth outer fabric surface that lowers particle cling and build-up in cleanroom applications. Alliance products have a unique anti-static system that assures static dissipation in sensitive cleanroom applications, while providing a liquid-proof barrier that ensures protection from hazardous substances used in Biotech and Pharmaceutical cleanrooms.
Packaging: 100 per case. Packed in poly-lined bag within brown corrugated box.