Monarch® DNA Capture Beads, New England Biolabs
Supplier: New England Biolabs (NEB)
T3005L
76470-762EA 77 USD
76470-762
Monarch® DNA Capture Beads, New England Biolabs
Beads Glass Beads
These specialized glass beads are optimized for capturing HMW DNA in the Monarch HMW DNA workflows.
  • A component of the Monarch HMW DNA extraction kits
  • Borosilicate glass beads of 4 mm
  • Optimized for extraction of HMW DNA
  • Smooth surface to maximize elution efficiency
  • Easy to handle when coupled with the Monarch bead retainers