LAMP Fluorescent Dye, New England Biolabs
Supplier: New England Biolabs (NEB)
B1700S
76470-738EA 54 USD
76470-738
LAMP Fluorescent Dye, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The LAMP fluorescent dye is an intercalating dye that can be used to track a LAMP reaction in real-time by fluorescence measurement.
  • Follow a LAMP reaction in real-time by fluorescence readout
It is provided at 50X concentration and is sufficient for at least 500 LAMP reactions (0.5 µl dye per 25 µl LAMP reaction). The SYBR®/FAM channel of common real-time fluorimeters can be used to read the dye.