Benchtop Centrifuges, Megafuge ST1 and ST1R Plus and Packages, Thermo Scientific
Supplier: Thermo Fisher Scientific
75009230 75009271 75016002 75016003 75016010 75017002 75017003 75016026US
76470-414EA 8300 USD
76470-414 76470-418 76470-426 76470-428 76470-430 76470-438 76470-442 76470-454
Benchtop Centrifuges, Megafuge ST1 and ST1R Plus and Packages, Thermo Scientific
Centrifuges Benchtop Centrifuges
The Thermo Scientific™ Megafuge ST1/ST1R Plus benchtop centrifuges offer intuitive controls and versatility for power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life. Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.
Compact benchtop dimensions optimize working area, the ergonomic height and curved design with motorized lid latch ensure easy sample loading and unloading, easy cleaning and promotes user safety.
System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Certifications: IVD certified.
Ordering information: Centrifuge only, optional Cell Culture and Blood Tube packages are also available.
Cell Culture package includes TX400 rotor, set of four 400 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 4×50 ml conical tube adapters, set of four 9×15 ml conical tube adapters.
Blood Tube package includes TX400 rotor, set of four 400ml buckets, set of four ClickSeal™ biocontainment lids, set of four 19×5/7 ml blood tube and 14×10 ml blood tube adapters.