VWR® Micro Weigh Boat
Supplier: VWR International
120836V
76470-181CS 160.68 USD
76470-181
VWR® Micro Weigh Boat
Boats Weighing Boats
Ideal for microbalance weighing and/or distribution of very small amounts of sampling material.
  • Inert PS material enables the boat and contents to be added to solutions to aid 100% sample recovery, where protocols allow
  • Anti-static properties prevent sample retention from static electricity
  • Large tabs make the boats easy to use, position and hold, especially with forceps
The VWR® Micro Weigh Boat is designed with anti-static properties for the weighing and distribution of small amounts of materials. The tabs allow for the weigh boat to be handled with forceps or other small instruments for placing the contents into flasks or other containers. Holds approximately 500 µl.