MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
Supplier: ELECTRON MICROSCOPY SCIENCE
80074-BAS-SS
76468-748EA 520.39 USD
76468-748
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
Standards Color Reference and Spectroscopy Standards
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 µ diamond finish and carbon coated. Supplied in 2, 3, and 5 mm diameter and a location map for standard identification. They are supplied with fully authenticated certificates of analysis.