Kleenguard™ A40 Liquid and Particle Protection Shoe Covers, Kimberly-Clark Professional
Supplier: Ansell Healthcare
27000
76468-270CS 321.87 USD
76468-270
Kleenguard™ A40 Liquid and Particle Protection Shoe Covers, Kimberly-Clark Professional
Disposable Garments
Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates, the KleenGuard™ A40 material provides employees a better liquid barrier and a better particulate barrier than leading competitors.
  • Kleenguard™ A40 Liquid and Particle Protection Shoe Covers have an elastic opening, serged seams, and a seamless sole
  • Film laminate material offers better liquid and particulate barrier than leading competitors
  • Made to keep out debris, dry particulates, and liquid splashes
  • Kleenguard™ A40 protective garments pass NFPA 99 criteria for anti-static materials
Kleenguard™ Personal Protective Equipment (PPE) enables rather than inhibits, allowing for maximum productivity and safety. Keeping workers comfortable while offering effective protective clothing is a must, and Kleenguard™ A40 Liquid and Particle Protection Shoe Covers are a great solution. They feature serged seams, an elastic opening, and a seamless sole. They are 7" tall and one size fits all. Popular industries using this product include aviation, pharmaceutical manufacturing, wind blade manufacturing, liquid handling, general manufacturing, and so much more.
Certifications: Shoe covers pass NFPA 99 for antistatic materials, and pass ASTM F1670 testing for penetration of blood and bodily fluids.