Anti-EMD Mouse Monoclonal Antibody [clone: 5A10] (DyL488)

Supplier: Boster Biological Technology
M00714-DYL488
76467-664EA 564.28 USD
76467-664
Anti-EMD Mouse Monoclonal Antibody [clone: 5A10] (DyL488)
Antibodies
Mouse IgG Monoclonal Anti-Human Emerin Antibody DyLight®, 488 Conjugated, Flow Validated.

Type: Primary
Antigen: EMD
Clonality: Monoclonal
Clone: 5A10
Conjugation: DyL488
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR