Mouse Plexin C1 PicoKine; ELISA Kit, Boster
Supplier: Boster Biological Technology
EK1829
76466-644EA 819.21 USD
76466-644
Mouse Plexin C1 PicoKine; ELISA Kit, Boster
Assays ELISAs
Sandwich high sensitivity ELISA kit for quantitative detection of mouse Plexin C1. 96 wells/kit, with removable strips.