Anti-DHX9 Rabbit Polyclonal Antibody

Supplier: Boster Biological Technology
A02550-1
76464-932EA 407.19 USD
76464-932
Anti-DHX9 Rabbit Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for RNA Helicase A/DHX9 detection. Tested with WB, IHC-P, ICC/IF, IF, FCM, Direct ELISA in Human,Mouse,Rat.

Type: Primary
Antigen: DHX9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR