Anti-POLR2A Rabbit Polyclonal Antibody (For POLR2A detection)

Supplier: Boster Biological Technology
A01029
76464-104EA 407.19 USD
76464-104
Anti-POLR2A Rabbit Polyclonal Antibody (For POLR2A detection)
Antibodies
Rabbit IgG polyclonal antibody for POLR2A detection. Tested with IHC-P, Direct ELISA in Human,Mouse,Rat.

Type: Primary
Antigen: POLR2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR