Super Erecta® SiteSelect® Industrial Wire Shelving Posts, Metroseal Gray Epoxy, Metro™
Supplier: Metro
13PK4 13UPK4 27PK4 27UPK4 33PK4 33UPK4 54PK4 54UPK4 63PK4 63UPK4 70UPK4 74PK4 74UPK4 86PK4 86UPK4
76463-194EA 29.17 USD
76463-194 76463-196 76463-198 76463-200 76463-202 76463-204 76463-206 76463-208 76463-210 76463-212 76463-214 76463-216 76463-218 76463-220 76463-222
Super Erecta® SiteSelect® Industrial Wire Shelving Posts, Metroseal Gray Epoxy, Metro™
Furniture Shelves
SiteSelect® posts provide a visual guide for positioning and adjusting shelves.
  • Posts are available in heights on 12, 27, 33, 54, 63, 74 and 86"
  • Both stationary and mobile-ready configurations
  • Mobile-ready posts come in 70" height
  • An easy-to-identify double groove every 8" offers a quick way to align all shelves
  • Mobile posts come without leveling bolt assembly to accommodate stem casters
  • Stationary posts are equipped with a leveling bolt to adjust for uneven floors
SiteSelect® Posts are finished with Super Erecta® Metroseal Gray epoxy which has Microban® antimicrobial protection built in. Metroseal Gray resists against corrosion and is NSF approved to be used in wet environments.
All Metroseal posts have a 12-year warranty for corrosion. Microban® prevents the growth of microorganisms, such as mold, that causes damage and odor.
Ordering information: Shelves sold separately.