Super Erecta® Wall Mount Shelving Intermediate Post Bracket
Supplier: Metro
BCSK4
76463-106EA 16.92 USD
76463-106
Super Erecta® Wall Mount Shelving Intermediate Post Bracket
Furniture Shelves
Replacement intermediate bracket for post-type mount wall shelving.
  • 33" posts require 1 intermediate bracket
  • 54 and 63" posts require 2 intermediate brackets
Corrosion resistant Metroseal Gray epoxy finish features Microban® antimicrobial production protection to keep the product 'cleaner between cleanings'.