20 L Bead Bath with 15 L Lab Armor Beads, Cascade Sciences
Supplier: CASCADE SCIENCES, LLC.
CS-BB20
76462-640EA 3768.19 USD
76462-640
20 L Bead Bath with 15 L Lab Armor Beads, Cascade Sciences
Baths Bead Baths
Upgrade laboratory setup with a 20 L bead bath and 15 L of thermal beads.
  • Temperature range between –80 to +180 °C
  • No water
  • High thermal efficiency
  • Microbial/biofilm resistant
  • No warm up
Waterless laboratory bead baths reduce the contamination and messy condensation you see in water baths. Bead baths don't evaporate which improves temperature performance and saves energy.