Diversey™ Divosan® Plus Broad Spectrum Sanitizer
Supplier: Janitorial Supplies
DVO57571100
76462-020EA 294.58 USD
76462-020
Diversey™ Divosan® Plus Broad Spectrum Sanitizer
Cleaning Agents Detergents and Surface Cleansers
Divosan® Plus is a peracetic acid based, broad spectrum sanitizer. It is suitable for use in dairies, breweries, soft drink and all other food processing operations. Divosan Plus is completely non-foaming and sanitizes effectively at temperatures as low as 40 °F.
  • Completely non-foaming and suitable for clean-in-place (CIP) and spray applications
  • Sanitizes and reduces bacteria in 1 minute, controls spoilage organisms; controls molds and yeast
  • Effective at temperatures as low as 40 °F
Application: Beverage, Dairy and Food Processing Operations;
Applicable Material: Aluminum; Stainless Steel; Mild Steel; Brass; Copper; Galvanized Surfaces;
Chemical Compound: Acetic Acid; Hydrogen Peroxide; Peracetic Acid;
Dirt Types: Bacteria; Mold; Spoilage Organisms; Yeast.