Diversey™ Avert Sporicidal Disinfectant Cleaner Chlorine Wipes, Essendant
Supplier: Janitorial Supplies
DVO100895790 DVO100895931
76461-976CT 561.35 USD
76461-976 76461-978
Diversey™ Avert Sporicidal Disinfectant Cleaner Chlorine Wipes, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Avert™ Sporicidal Disinfectant Cleaner Wipes clean, disinfect, and deodorize hard, non-porous surfaces in one labor-saving step with no rinsing required. Designed for use on hard, nonporous surfaces such as those found in hospitals and other healthcare facilities.
  • Effective in four minutes against Clostridium difficile (C. diff) spores
  • Heavy-duty disinfectant wipes clean, disinfect, and deodorize in one step
  • Bleach-based formula means no quat-binding concerns
  • No added fragrance or dyes
Avert Sporicidal Disinfectant Cleaner Chlorine Wipes are designed to clean and disinfect sinks, faucets, fixtures, bathrooms, restrooms, tubs, and showers.
Use these single-ply wipes on hard, non-porous surfaces to eliminate Clostridium Difficile dirt types.