D-(-)-Sorbitol

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02194742.5
76461-504EA 59.54 USD
76461-504
D-(-)-Sorbitol
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 94…98 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR