Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219455794
76461-488EA 725.96 USD
76461-488
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Formula: NH₂C(CH₂OH)₃
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 169…172 °C
Density: 1.328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR