G418 disulfate

Supplier: 0000012207
Warning

0215878294
76461-468EA 616.31 USD
76461-468
G418 disulfate
G418 disulphate
Formula: C₂₀H₄₄N₄O₁₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR