Low-Smoke, Zero-Halogen 16 to 10 Gauge Wire Marking Sleeves, 3" Core, Brady
Supplier: Brady Worldwide
2LSZH-187-2-YL-2
76461-348RL 2373.36 USD
76461-348
Low-Smoke, Zero-Halogen 16 to 10 Gauge Wire Marking Sleeves, 3" Core, Brady
Labels
The B-7641 Heat Shrink Sleeve from Brady® is a low-smoke, halogen-free identification material used for wire and cable identification.
  • Flame-retardant, heat-shrinkable
  • Low-smoke, zero-halogen
  • Double-side printable
The B-7641 Heat Shrink Sleeve is made of halogen-free, flexible flame-retardant, heat-shrinkable polyolefin tubing, offering fire safety properties and minimal smoke emission. Compatible with the most stringent requirements in the market today to enhance fire safety needs. Sleeves offer a 2:1 heat-shrink ratio.
Sleeves are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®72 Label Printer, BradyPrinter i7100 Industrial Label Printer, THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 76251-721 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 76460-980 for a different printer.