Polyester Chemical-Resistant Slide Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-164-481-3
76461-108RL 355.49 USD
76461-108
Polyester Chemical-Resistant Slide Labels, 3" Core, Brady
Labels
Super Chemical Resistant Polyester (B-481) is designed for use in the slide staining process and other laboratory identifications which require superior chemical resistance.
  • Autoclavable
The B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers from Brady®. B-481 performs well in the commonly used H & E (hematoxilin and eosin) slide staining process. Excellent resistance to a wide variety of chemicals, oils and water. Service temperature of ‒80 to +130 °C (‒112 to +266 °F).
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 490006-116 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6406 for a different printer.