Matte Silver Polyester with Rubber Adhesive Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-19-480-1
76461-104RL 317.61 USD
76461-104
Matte Silver Polyester with Rubber Adhesive Labels, 3" Core, Brady
Labels
This B-480 material is a thermal transfer printable polyester, with a rubber based adhesive that is ideal for oily or greasy applications.
  • Metallic appearance
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 76244-722 for the BradyPrinter i5100 or BRADY IP® Series LabelMore Product Information