Continuous Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-106-486
76461-014RL 412.33 USD
76461-014
Continuous Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Labels
Ultra Aggressive Metallized Polyester (B-486B) is ideal for rating/serial plates that use barcodes, alphanumerics, graphic symbols, and logos, and require nameplate-like quality.
  • Aggressive adhesive
  • Heat-resistant
  • Metallic
  • Permanent adhesive
Ultra Aggressive Metallized Polyester (B-486B) is a matte, light gray material that has a permanent rubber-based adhesive and is thermal transfer printable. Designed for high adhesion to textured metals, powder-coated surfaces, and low energy plastics. Service temperature of ‒40 to +120 °C (‒40 to +248 °F).
Labels are designed for use with BBP®81 Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Certifications: UL approved. CSA accepted.
Ordering information: A ribbon is required to print this material. Use Part Number 89077-374 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 94000-380 for a different printer.