VWR® pHenomenal® CO 3100 H Conductivity/TDS/°C Meter, Handheld
Supplier: VWR International
76460-508 76460-530
76460-508EA 1106.25 USD
76460-508 76460-530
VWR® pHenomenal® CO 3100 H Conductivity/TDS/°C Meter, Handheld
Electrochemistry Meters Conductivity Meters
This user friendly portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applications. Complete with waterproof USB output for connection to a computer. GLP compliant.
  • Large LCD graphic display with continuous LED backlight showing conductivity, TDS and temperature values simultaneously
  • Data logging function selectable between 1 min and 1 h
  • 5000 data set memory capacity with automatic storing; additional 200 data set memory for manual storage
  • Automatic power shut off: Adjustable from 10 min to 24 h
  • Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when re-calibration is required
  • Low energy consumption: 4× AA batteries for over 1000 operating hours
Ordering information: Conductivity probes for low conductivities are available on request. Please contact your local Avantor sales office for further information.
Delivery information: Ready to use portable kit CO 3100 H (76460-508) includes meter in robust plastic carrying case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml conductivity standard 0.01 mol/L KCl and conductivity probe with built-in temperature sensor CO 11 (1.5 m cable).
76460-530 includes meter, case, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual.