VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld

Supplier: VWR International
76460-504 76460-518
76460-504EA 799.75 USD
76460-504 76460-518
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Electrochemistry Meters pH and Multi-Parameter Meters
This user friendly, portable instrument with robust IP 67 rated housing provides high resolution and accuracy for precise measurements in field applications. Complete with waterproof USB output for connection to a computer. GLP compliant.

  • Large LCD graphic display with continuous LED backlight shows both pH/mV and temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h
  • 5000 data set memory capacity with automatic storing; additional 200-point memory for manual storage
  • Automatic power shutoff: Adjustable from 10 min to 24 h
  • Automatic buffer recognition of DIN and NIST buffers (1,68/4,00/6,86/9,18/12,54)
  • Three additional technical buffers at 25 °C (4,00/7,00/10,00) in memory (pH 10,00 not included in delivery)
  • Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
  • Low energy consumption: 4× AA batteries for over 1000 operating hours

Delivery information: Ready to use portable pH kit pH 1100 H (76460-504) includes instrument in robust plastic carrying case, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, gel epoxy pH electrode with built-in temperature sensor 111 (1 m cable).
76460-518 includes meter, case, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR