VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/conductivity/oxygen), Bench

Supplier: VWR International
76460-494 76460-496 76460-520
76460-494EA 2104.91 USD
76460-494 76460-496 76460-520
VWR® pHenomenal® MU 6100 L Multi-Parameter Meter (pH/conductivity/oxygen), Bench
Electrochemistry Meters pH and Multi-Parameter Meters
This user friendly instrument, with articulated electrode stand and IP 43-rated housing, provides high resolution and accuracy for precise multi-parameter measurements. This meter can either measure pH/mV and conductivity or pH/mV and oxygen simultaneously. Complete with USB output for connection to a computer. GLP compliant.

  • Large graphic LCD with continuous LED backlight shows the measured parameter and the temperature value simultaneously
  • Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets
  • Automatic power shut off: Adjustable from 10 min to 24 h
  • Automatic buffer recognition of DIN and NIST buffers (1,68/4,00/6,86/9,18/12,54)
  • Three additional technical buffers at 25 °C (4,00/7,00/10,00) in memory
  • Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
  • Low energy consumption. Choice of mains supply or 4× AA batteries for over 1000 operating hours

Ordering information: Conductivity probes for low conductivities are available on request. Please contact your local Avantor sales office for further information.

Delivery information: Ready to use MU 6100 L S1 kit (76460-494 ) includes bench multi-parameter meter, glass pH electrode pHenomenal 221 with built-in temperature sensor (1 m cable), conductivity probe CO 11 with built-in temperature sensor (1,5 m cable), articulated stand, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, short instructions and instruction manual (22 languages) on CD ROM.
The MU 6100 L S2 kit (76460-496) includes meter, stand, 4 LR06 batteries, short instructions, CD ROM with manual, 50 ml plastic beaker, 50 ml buffer 4, 50 ml buffer 7, plugin power supply, pH electrode pHenomenal 221, 50 ml conductivity standard 0.01 mol/L KCl, conductivity cell CO 11, 30 ml cleaning solution, 50 ml electrolyte, 3 exchange caps, DO electrode OXY 11 (3 m cable).
76460-520 includes meter, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual and plugin power supply.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR